• USA Home
  • Search Results

  • trimethylsilylcyanide9921767724911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethylsilyl cyanide

2 Products
Synonym:   Cyanotrimethylsilane, TMSCN
Image of Trimethylsilyl cyanide
CAS Number:
Linear Formula:
 (CH3)3SiCN
Molecular Weight:
 99.21
 Beilstein/REAXYS Number:
 1737612
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More