• USA Home
  • Search Results

  • trimethylsilyldiazomethanesolution114221810718111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Trimethylsilyl)diazomethane solution

2 Products
Synonym:   (Diazomethyl)trimethylsilane
Image of (Trimethylsilyl)<WBR>diazomethane solution
CAS Number:
Linear Formula:
 (CH3)3SiCHN2
Molecular Weight:
 114.22
 Beilstein/REAXYS Number:
 1902903
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More