• USA Home
  • Search Results

  • trimethyltinchloridesolution19927106645111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethyltin chloride solution

2 Products
Image of Trimethyltin chloride solution
CAS Number:
Linear Formula:
 (CH3)3SnCl
Molecular Weight:
 199.27
 Beilstein/REAXYS Number:
 3535111
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More