• USA Home
  • Search Results

  • trimipraminemaleatesalt4105152178811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimipramine maleate salt

1 Products
Image of Trimipramine maleate salt
CAS Number:
Empirical Formula (Hill Notation):
 C20H26N2 · C4H4O4
Molecular Weight:
 410.51
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More