• USA Home
  • Search Results

  • trinbutylphosphate2663112673811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tri-n-butyl phosphate

2 Products
Synonym:   TBP, TBPA, Tributyl phosphate
Image of Tri-n-butyl phosphate
CAS Number:
Linear Formula:
 (CH3(CH2)3O)3PO
Molecular Weight:
 266.31
 Beilstein/REAXYS Number:
 1710584
 EC Index Number:
 204-800-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
Show More