• USA Home
  • Search Results

  • trioctylphosphine37064473153711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trioctylphosphine

2 Products
Synonym:   P(Oct)3, TOP
Image of Trioctylphosphine
CAS Number:
Linear Formula:
 [CH3(CH2)7]3P
Molecular Weight:
 370.64
 Beilstein/REAXYS Number:
 1776995
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Functional group
  Assay
Show More