• USA Home
  • Search Results

  • triphenylmethanethiol27640369577011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylmethanethiol

1 Products
Synonym:   Triphenylmethyl mercaptan, Trityl mercaptan
Image of Triphenylmethanethiol
CAS Number:
Linear Formula:
 (C6H5)3CSH
Molecular Weight:
 276.40
 Beilstein/REAXYS Number:
 2113168
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More