• USA Home
  • Search Results

  • triphenylmethylamine25934582440811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylmethylamine

1 Products
Synonym:   Tritylamine
Image of Triphenylmethylamine
CAS Number:
Linear Formula:
 (C6H5)3CNH2
Molecular Weight:
 259.34
 Beilstein/REAXYS Number:
 2113674
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
325430 Triphenylmethylamine, 99%,