• USA Home
  • Search Results

  • triphenylphosphate3262811586611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenyl phosphate

4 Products
Image of Triphenyl phosphate
CAS Number:
Linear Formula:
 (C6H5O)3PO
Molecular Weight:
 326.28
 Beilstein/REAXYS Number:
 1888236
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Potency
  Application(s)
Show More