• USA Home
  • Search Results

  • triphenylphosphine2622960335011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylphosphine

3 Products
Synonym:   Phosphorustriphenyl
Image of Triphenylphosphine
CAS Number:
Linear Formula:
 (C6H5)3P
Molecular Weight:
 262.29
 Beilstein/REAXYS Number:
 610776
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Product Line
  Potency
  Reaction Suitability
  Functional group
  Solubility
  Melting Point
  Assay
Show More