• USA Home
  • Search Results

  • triphenylphosphinehydrobromide34320639981111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylphosphine hydrobromide

1 Products
Synonym:   Ph3P · HBr, Ph3P · HBr, Triphenylphosphonium bromide
Image of Triphenylphosphine hydrobromide
CAS Number:
Linear Formula:
 (C6H5)3P · HBr
Molecular Weight:
 343.20
 Beilstein/REAXYS Number:
 3633387
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Assay
Show More