• USA Home
  • Search Results

  • triphenylphosphinepolymerbound262293931911411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylphosphine, polymer-bound

4 Products
Synonym:   Copolymer of styrene and divinylbenzene, diphenylphosphinated, Diphenylphosphino-polystyrene, Polystyrene crosslinked with divinylbenzene, diphenylphosphinated
Image of Triphenylphosphine, polymer-bound
CAS Number:
Linear Formula:
 -C6H4P(C6H5)2
Molecular Weight:
 262.29
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Extent of labeling
  Particle Dimension
Show More