• USA Home
  • Search Results

  • triphenylphosphoranylideneacetaldehyde30432213675611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Triphenylphosphoranylidene)acetaldehyde

1 Products
Synonym:   (Formylmethylene)triphenylphosphorane
Image of (Triphenylphosphoranylidene)<WBR>acetaldehyde
CAS Number:
Linear Formula:
 HCOCH=P(C6H5)3
Molecular Weight:
 304.32
 Beilstein/REAXYS Number:
 523797
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More