• USA Home
  • Search Results

  • triphenylsilanol2764079131111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triphenylsilanol

1 Products
Synonym:   Hydroxytriphenylsilane
Image of Triphenylsilanol
CAS Number:
Linear Formula:
 (C6H5)3SiOH
Molecular Weight:
 276.40
 Beilstein/REAXYS Number:
 985007
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
143723 Triphenylsilanol, 98%,