• USA Home
  • Search Results

  • triprolidinehydrochloride31485613879011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triprolidine hydrochloride

2 Products
Synonym:   (E)-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride, trans-2-[3-(1-Pyrrolidinyl)-1-p-tolylpropenyl]pyridine hydrochloride
Image of Triprolidine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H22N2 · HCl
Molecular Weight:
 314.85
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More