• USA Home
  • Search Results

  • tripropionin2602813945711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tripropionin

1 Products
Synonym:   Glyceryl tripropionate
Image of Tripropionin
CAS Number:
Linear Formula:
 (C2H5CO2CH2)2CH(O2CC2H5)
Molecular Weight:
 260.28
 Council of Europe no.:
 10657
 EC Number:
 FEMA Number:
 3286
 Flavis number:
 9.263
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Special Grade
  Organoleptic property
  Titertest
  Biological Source
  Assay
Show More