• USA Home
  • Search Results

  • triptolylphosphine30437103895511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tri(p-tolyl)phosphine

1 Products
Synonym:   TPTP
Image of Tri(<I>p</I>-tolyl)phosphine
CAS Number:
Linear Formula:
 (CH3C6H4)3P
Molecular Weight:
 304.37
 Beilstein/REAXYS Number:
 651045
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More