• USA Home
  • Search Results

  • tris24ditertbutylphenylphosphite646923157004411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(2,4-di-tert-butylphenyl) phosphite

1 Products
Synonym:   2,4-Bis(1,1-dimethylethyl)phenol phosphite (3:1), Tri(2,4-di-tert -butylphenyl) phosphite, Tri(2,4-di-t -butylphenyl) phosphite
Image of Tris(2,4-di-<I>tert</I>-butylphenyl) phosphite
CAS Number:
Linear Formula:
 [[(CH3)3C]2C6H3O]3P
Molecular Weight:
 646.92
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More