• USA Home
  • Search Results

  • tris2butoxyethylphosphate398477851311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(2-butoxyethyl) phosphate

2 Products
Synonym:   Phosphoric acid tris(2-butoxyethyl) ester
Image of Tris(2-butoxyethyl) phosphate
CAS Number:
Linear Formula:
 [CH3(CH2)3OCH2CH2O]3P(O)
Molecular Weight:
 398.47
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More