• USA Home
  • Search Results

  • tris2chloroethylphosphite2694914008911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(2-chloroethyl) phosphite

1 Products
Image of Tris(2-chloroethyl) phosphite
CAS Number:
Linear Formula:
 (ClCH2CH2O)3P
Molecular Weight:
 269.49
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More