• USA Home
  • Search Results

  • tris2phenylpyridinatoc2niridiumiii654789492886611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris[2-phenylpyridinato-C2,N]iridium(III)

2 Products
Synonym:   Ir(ppy)3, Iridium, tris[2-(2-pyridinyl-κN)phenyl-κC]
Image of Tris[2-<WBR>phenylpyridinato-<WBR>C<SUP>2</SUP>,<I>N</I>]iridium(III)
CAS Number:
Empirical Formula (Hill Notation):
 C33H24IrN3
Molecular Weight:
 654.78
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  OLED Device Performance
  Fluorescence Test
  Orbital energy
Show More