• USA Home
  • Search Results

  • tris4heptadecafluorooctylphenylphosphine15164428447292011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris[4-(heptadecafluorooctyl)phenyl]phosphine

1 Products
Synonym:   Tris[4-(perfluorooctyl)phenyl]phosphine
Image of Tris[4-<WBR>(heptadecafluorooctyl)<WBR>phenyl]<WBR>phosphine
CAS Number:
Empirical Formula (Hill Notation):
 C42H12F51P
Molecular Weight:
 1516.44
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More