• USA Home
  • Search Results

  • tris8hydroxyquinolinealuminum45943208533811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris-(8-hydroxyquinoline)aluminum

3 Products
Synonym:   8-Hydroxyquinoline aluminum salt, Alq3, Aluminum 8-hydroxyquinolinate, Aluminum oxinate, Tris-(8-hydroxyquinolinato)aluminum
Image of Tris-<WBR>(8-<WBR>hydroxyquinoline)<WBR>aluminum
CAS Number:
Empirical Formula (Hill Notation):
 C27H18AlN3O3
Molecular Weight:
 459.43
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  OLED Device Performance
  Assay
  Orbital energy
Show More