• USA Home
  • Search Results

  • trisdibenzylideneacetonedipalladium0915725136451311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(dibenzylideneacetone)dipalladium(0)

2 Products
Synonym:   Pd2dba3, Pd2(dba)3
Image of Tris(dibenzylideneacetone)<WBR>dipalladium(0)
CAS Number:
Linear Formula:
 (C6H5CH=CHCOCH=CHC6H5)3Pd2
Molecular Weight:
 915.72
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More