• USA Home
  • Search Results

  • trisdiethylaminophosphine24736228311611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(diethylamino)phosphine

1 Products
Synonym:   Hexaethylphosphorous triamide, Hexaethyltriamidophosphite, Hexaethyltriaminophosphine, Phosphorous acid tris(diethylamide), Tris(N,N-diethylamino)phosphine, Tris(diethylamido)phosphine
Image of Tris(diethylamino)<WBR>phosphine
CAS Number:
Linear Formula:
 P[N(C2H5)2]3
Molecular Weight:
 247.36
 Beilstein Registry Number:
 636187
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More