• USA Home
  • Search Results

  • trisethylmethylamidotertbutylimidotantalumv4263851129299211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(ethylmethylamido)(tert-butylimido)tantalum(V)

1 Products
Image of Tris(ethylmethylamido)<WBR>(<I>tert</I>-<WBR>butylimido)<WBR>tantalum(V)
CAS Number:
Empirical Formula (Hill Notation):
 C13H33N4Ta
Molecular Weight:
 426.38
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More