• USA Home
  • Search Results

  • trisodiumcitratedihydrate29410613204311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

tri-Sodium citrate dihydrate

5 Products
Synonym:   Citric acid trisodium salt dihydrate, Sodium citrate tribasic dihydrate, Trisodium Citrate, Sodium Citrate, Dihydrate, Trisodium citrate dihydrate
Image of tri-Sodium citrate dihydrate
CAS Number:
Linear Formula:
 HOC(COONa)(CH2COONa)2 · 2H2O
Molecular Weight:
 294.10
 Beilstein/REAXYS Number:
 6104939
 EC Index Number:
 200-675-3
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Product Line
  Assay
  Physical Form
  Potency
  Melting Point
  Bulk Density
  pH Value
Show More