• USA Home
  • Search Results

  • tristriphenylphosphinerhodiumichloride925221469495211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(triphenylphosphine)rhodium(I) chloride

2 Products
Synonym:   NSC 124140, RhCl(PPh3)3, Rhodium(I) tris(triphenylphosphine) chloride, Wilkinson’s catalyst
Image of Tris(triphenylphosphine)<WBR>rhodium(I) chloride
CAS Number:
Linear Formula:
 [(C6H5)3P]3RhCl
Molecular Weight:
 925.22
 Beilstein/REAXYS Number:
 4581440
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More