• USA Home
  • Search Results

  • tristriphenylphosphinerutheniumiidichloride958831552949411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tris(triphenylphosphine)ruthenium(II) dichloride

1 Products
Synonym:   Dichlorotris(triphenylphosphine)ruthenium(II), Ruthenium(II)-tris(triphenylphosphine) dichloride, Tris(triphenylphosphine)dichlororuthenium
Image of Tris(triphenylphosphine)<WBR>ruthenium(II) dichloride
CAS Number:
Linear Formula:
 [(C6H5)3P]3RuCl2
Molecular Weight:
 958.83
 Beilstein/REAXYS Number:
 4935939
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More