• USA Home
  • Search Results

  • tritonx100reduced123459204634911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triton X-100 reduced

2 Products
Synonym:   Polyoxyethylene (10) isooctylcyclohexyl ether
Image of Triton<SUP>™</SUP> X-100 reduced
CAS Number:
Linear Formula:
 4-(C8H17)C6H10(OCH2CH2)nOH, n~10
 MDL number:
 NACRES:
 NA.25
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More