• USA Home
  • Search Results

  • tritonx100solution12345900293111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triton X-100 solution

1 Products
Synonym:   4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether, Triton X-100
Image of Triton<SUP>™</SUP> X-100 solution
CAS Number:
Linear Formula:
 t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
 Beilstein/REAXYS Number:
 2315025
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  ID clarifier
  Density
Show More