• USA Home
  • Search Results

  • tritonx4512345900293111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triton X-45

1 Products
Synonym:   4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol 4-tert-octylphenyl ether
Image of Triton<SUP>™</SUP> X-45
CAS Number:
Linear Formula:
 t-Oct-C6H4-(OCH2CH2)xOH, x= ~5
 Beilstein/REAXYS Number:
 2315025
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
X45 Triton X-45,