• USA Home
  • Search Results

  • trolamine1491910271611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trolamine

2 Products
Synonym:   2,2′,2′′-Nitrilotriethanol, Triethanolamine, Tris(2-hydroxyethyl)amine
Image of Trolamine
CAS Number:
Linear Formula:
 (HOCH2CH2)3N
Molecular Weight:
 149.19
 Beilstein/REAXYS Number:
 1699263
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
Show More