• USA Home
  • Search Results

  • trolamineimpurityb1051411142211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trolamine impurity B

1 Products
Synonym:   2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine, Diethanolamine
Image of Trolamine impurity B
CAS Number:
Linear Formula:
 HN(CH2CH2OH)2
Molecular Weight:
 105.14
 Beilstein/REAXYS Number:
 605315
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More