• USA Home
  • Search Results

  • trometamol121147786111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trometamol

1 Products
Synonym:   Tris, Tromethamine
Image of Trometamol
CAS Number:
Empirical Formula (Hill Notation):
 C4H11NO3
Molecular Weight:
 121.14
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More