• USA Home
  • Search Results

  • tropicamide28435150875411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tropicamide

4 Products
Synonym:   N-Ethyl-2-phenyl-N-(4-pyridylmethyl)hydracrylamide, Ro 1-7683
Image of Tropicamide
CAS Number:
Empirical Formula (Hill Notation):
 C17H20N2O2
Molecular Weight:
 284.35
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
  Solubility
Show More