• USA Home
  • Search Results

  • tropicamiderelatedcompoundd1361510382211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tropicamide Related Compound D

1 Products
Synonym:   α-Toluic acid, α-Tolylic acid, Benzeneacetic acid, PAA, Phenylacetic acid
Image of Tropicamide Related Compound D
CAS Number:
Linear Formula:
 C6H5CH2CO2H
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 1099647
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More