• USA Home
  • Search Results

  • trypanbluesolution960817257111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trypan Blue solution

2 Products
Image of Trypan Blue solution
CAS Number:
Empirical Formula (Hill Notation):
 C34H24N6Na4O14S4
Molecular Weight:
 960.81
 Beilstein Registry Number:
 4360496
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
Show More