• USA Home
  • Search Results

  • trypsinedtasolution1x1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trypsin-EDTA Solution 1X

3 Products
Synonym:   Trypsin-EDTA solution
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Culture media components
  Sterilization
  Ice Shipping
  ID clarifier
Show More