• USA Home
  • Search Results

  • trypsinfrombovinepancreas12345900207711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trypsin from bovine pancreas

8 Products
Image of Trypsin from bovine pancreas
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.4.21.4  ( BRENDA | IUBMB )
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Suitability
  Type
  Quality clarifier
  Ice Shipping
  Chemical Composition
  Physical Form
Show More

Product #
Description
T1426 Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein,
T8003 Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein,
T8658 Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder,
T9201 Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid,
T1005 Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein,
T9935 Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture,
T8802 Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein,
T7309 Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications,