• USA Home
  • Search Results

  • trypsininhibitorfromchickeneggwhite12345903581811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trypsin inhibitor from chicken egg white

2 Products
Synonym:   Ovomucoid
Image of Trypsin inhibitor from chicken egg white
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.77
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Type
Show More