• USA Home
  • Search Results

  • trypsinsolutionfromporcinepancreas12345900207711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trypsin solution from porcine pancreas

3 Products
CAS Number:
 Enzyme Commission (EC) Number:
 3.4.21.4  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Physical Form
  pH Value
  Application(s)
  Sterilization
  Product Line
  Molecular weight
  Ice Shipping
Show More