• USA Home
  • Search Results

  • tryptamine160226154111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tryptamine

2 Products
Synonym:   2-(3-Indolyl)ethylamine, 3-(2-Aminoethyl)indole
Image of Tryptamine
CAS Number:
Empirical Formula (Hill Notation):
 C10H12N2
Molecular Weight:
 160.22
 Beilstein/REAXYS Number:
 125513
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More