• USA Home
  • Search Results

  • tryptaminehydrochloride1966834394211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tryptamine hydrochloride

1 Products
Synonym:   2-(3-Indolyl)ethylamine hydrochloride, 3-(2-Aminoethyl)indole hydrochloride
Image of Tryptamine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C10H12N2 · HCl
Molecular Weight:
 196.68
 Beilstein/REAXYS Number:
 3568419
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More