• USA Home
  • Search Results

  • tskgelvmpak25hplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

TSKgel® VMpak-25 HPLC Column

2 Products
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Operating parameter
  Chromatography Mode
  pH Range
  Titertest
  Particle Dimension
  Matrix active group
  Material
  Manufacturer designation
  Composite Dimension
Show More