• USA Home
  • Search Results

  • tulobuterolhydrochloride264195677601311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tulobuterol hydrochloride

1 Products
Synonym:   2-tert-Butylamino-1-(2-chlorophenyl)ethanol hydrochloride
Image of Tulobuterol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H18ClNO · HCl
Molecular Weight:
 264.19
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More