• USA Home
  • Search Results

  • tungstenhexacarbonyl351901404011011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tungsten hexacarbonyl

3 Products
Synonym:   Hexacarbonyltungsten, Tungsten carbonyl, Tungsten hexacarbonyl
Image of Tungsten hexacarbonyl
CAS Number:
Linear Formula:
 W(CO)6
Molecular Weight:
 351.90
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purification method
Show More