• USA Home
  • Search Results

  • tween2012345900564511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

TWEEN® 20

10 Products
Synonym:   Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
Image of TWEEN<SUP>®</SUP> 20
CAS Number:
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Product Line
  Molecular weight
  Special Grade
  Density
  Assay
  Chemical Composition
  Transition temperature
  HLB Number
  Critical micelle concentration
  Physical Form
  ID clarifier
Show More