• USA Home
  • Search Results

  • tween8012345900565611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tween 80

10 Products
Synonym:   Tween 80
Image of Tween<TMSYMBOL></TMSYMBOL> 80
CAS Number:
 EC Index Number
 500-019-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Suitability
  pH Value
  Vapor pressure
  Special Grade
  Miscellaneous Specifications
  HLB Number
  Critical micelle concentration
  Physical Form
  Transition temperature
  Molecular weight
  Chemical Composition
  Density
  ID clarifier
Show More