• USA Home
  • Search Results

  • typhaneoside7706910447268611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Typhaneoside

2 Products
Synonym:   3-[(O-6-Deoxy-α-L-mannopyranosyl-(1→2)-O-[6-deoxy-a-L-mannopyranosyl-(1→6)]-β-D-glucopyranosyl)oxy]-5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-1-benzopyran-4-one, Isorhamnetin 3-(2′′-rhamnosylrutinoside)
Image of Typhaneoside
CAS Number:
Empirical Formula (Hill Notation):
 C34H42O20
Molecular Weight:
 770.69
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
Show More